Anti-TNFR1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK109-401-308EA 1020 EUR
ROCK109-401-308
Anti-TNFR1 Rabbit Polyclonal Antibody
Antikörper
This antiserum against Human TNF p55 Receptor (sTNFRp55) has been tested for use in ELISA,  radioimmunoassays, immunoblotting and immunoprecipitation. Reactivity in other immunoassays is unknown.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: TNFR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Primate
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR