Anti-IL6 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK109-401-310EA 1020 EUR
ROCK109-401-310
Anti-IL6 Rabbit Polyclonal Antibody
Antikörper
IL-6 antisera has been tested for use in neutralizations, ELISA, immuno-histochemistry, radioimmunoassay, immunoprecipitation, and immunoblotting. Reactivity in other immunoassays is unknown.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:400 - 1:800; Immunoprecipitation: 1:400 - 1:800;

Type: Primary
Antigen: IL6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR