Anti-CXCL8 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK109-401-311EA 1020 EUR
ROCK109-401-311
Anti-CXCL8 Rabbit Polyclonal Antibody
Antikörper
IL 8 antibody has been tested for use in ELISA and recognizes IL-8 found in cells, supernatants and other body fluids.  Reactivity in other immunoassays is unknown.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000; Immunochemistry: 1:400 - 1:800; Immunoprecipitation: 1:400 - 1:800;

Type: Primary
Antigen: CXCL8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR