Anti-IL10 Rabbit Monoclonal Antibody (Unconjugated) [clone: 8G1]

Lieferant: Rockland Immunochemicals
ROCK109-401-312EA 1020 EUR
ROCK109-401-312
Anti-IL10 Rabbit Monoclonal Antibody (Unconjugated) [clone: 8G1]
Antikörper
This antiserum has been tested for use in neutralizations, ELISA, immunochemistry, radioimmunoassay, immunoprecipitation, immunochemistry and immunoblotting.  The antibody recognizes hrIL-10.  Reactivity in other immunoassays is unknown.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Primary
Antigen: IL10
Clonality: Monoclonal
Clone: 8G1
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR