Anti-CCL3 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK109-401-315EA 1020 EUR
ROCK109-401-315
Anti-CCL3 Rabbit Polyclonal Antibody
Antikörper
This antiserum against Human MIP-1a has been tested for use in ELISA of human MIP-1a found in cell supernatants or in plasma or serum.  Reactivity in other immunoassays is unknown.

Recommended Dilutions: ELISA: 1:1000 - 1:5000; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: CCL3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR