Anti-TF Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK109-4134EA 249 EUR
ROCK109-4134
Anti-TF Rabbit Polyclonal Antibody
Antikörper
This product has been assayed against 1.0 µg of Transferrin (Human Serum) in a standard ELISA using Peroxidase conjugated Affinity Purified anti-Rabbit IgG (H&L) (Goat) and ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid))

Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: TF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR