Anti-Mouse Serum Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK110-1101EA 482 EUR
ROCK110-1101
Anti-Mouse Serum Goat Polyclonal Antibody
Antikörper
Primary Goat Anti-MOUSE SERUM Reacts with Mouse

Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Mouse Serum
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR