Anti-IgG F(c) Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK110-1103EA 120 EUR
ROCK110-1103
Anti-IgG F(c) Goat Polyclonal Antibody
Antikörper
Secondary Goat Anti-IgG F(c) Reacts with Mouse

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR