Anti-Rabbit Serum Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK111-1101EA 249 EUR
ROCK111-1101
Anti-Rabbit Serum Goat Polyclonal Antibody
Antikörper
Anti-Rabbit serum antibody is suitable for western blotting, IP and for ELISA. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions:
Restore with deionized water (or equivalent); 2.0 ml

Type: Primary
Antigen: Rabbit Serum
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR