Anti-IgM µ Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK111-1107EA 120 EUR
ROCK111-1107
Anti-IgM µ Goat Polyclonal Antibody
Antikörper
Secondary Goat Anti-IgM mu chain Reacts with Rabbit (Lapine)

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: µ-chain
Host: Goat
Isotype: IgM
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR