Anti-IgG F(c) Donkey Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK111-7103EA 234 EUR
ROCK111-7103
Anti-IgG F(c) Donkey Polyclonal Antibody
Antikörper
Antibody Anti-Rabbit IgG F(c) is suitable for immunoblotting (western or dot blot), ELISA, and immunochemistry as well as other peroxidase-antibody based enzymatic assays requiring lot-to-lot consistency.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 2.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(c)
Host: Donkey
Isotype: IgG
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR