Anti-ATM Mouse Polyclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 10H11.E12]

Lieferant: Rockland Immunochemicals
ROCK200-303-400EA 498 EUR
ROCK200-303-400
Anti-ATM Mouse Polyclonal Antibody (HRP (Horseradish Peroxidase)) [clone: 10H11.E12]
Antikörper
This product has been tested by ELISA and western blotting against both the native and recombinant forms of the Protein. This reagent may also be suitable for immunoperoxidase electron microscopy and immunochemistry as well as other peroxidase-antibodies.

Recommended Dilutions: ELISA: 1:70,000; Western Blot: 1:600;
Restore with deionized water (or equivalent); 100 µl
Do not add Sodium Azide!

Type: Primary
Antigen: ATM
Clonality: Polyclonal
Clone: 10H11.E12
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Mouse, Rat, Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR