Anti-RFX5 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK200-401-191EA 379 EUR
ROCK200-401-191
Anti-RFX5 Rabbit Polyclonal Antibody
Antikörper
Primary Rabbit Anti-RFX5 (C-terminal) Reacts with Human, Mouse, Rat

Recommended Dilutions: ELISA: 1:5000 - 1:25000; Western Blot: 1:500 - 1:3000;

Type: Primary
Antigen: RFX5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: near the C-terminus
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR