Anti-IgG F(c) Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK210-1103EA 481 EUR
ROCK210-1103
Anti-IgG F(c) Goat Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG (Mouse) (mouse IgG)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR