Anti-IgG (H+L) Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK211-1102EA 424 EUR
ROCK211-1102
Anti-IgG (H+L) Goat Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG (Rabbit) (rabbit IgG)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: none
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR