Anti-IgG F(ab')2 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK305-4104EA 441 EUR
ROCK305-4104
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Goat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR