Anti-IgG (H+L) Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Rockland Immunochemicals
ROCK307-4302EA 293 EUR
ROCK307-4302
Anti-IgG (H+L) Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Golden Syrian Hamster
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR