Anti-ING5 Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK600-101-285EA 379 EUR
ROCK600-101-285
Anti-ING5 Goat Polyclonal Antibody
Antikörper
This affinity purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 28 kDa or 26 kDa in size corresponding to variants of ING5 by wester blotting.

Recommended Dilutions: ELISA: 1:4000 - 1:16000; Western Blot: 1:500 - 1:1000;

Type: Primary
Antigen: ING5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR