Anti-PPARD Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK600-401-420EA 783 EUR
ROCK600-401-420
Anti-PPARD Rabbit Polyclonal Antibody
Antikörper
This affinity purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a single band approximately 43 kDa in size corresponding to PPAR delta by western blot.

Recommended Dilutions: ELISA: 1:600,000; Western Blot: 1:500 - 1:5000;

Type: Primary
Antigen: PPARD
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: N-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Mouse, Rat, Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR