Anti-TGFB1 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK600-401-432EA 379 EUR
ROCK600-401-432
Anti-TGFB1 Rabbit Polyclonal Antibody
Antikörper
TGF beta 1 Antibody has been tested for use in ELISA and western blotting.  Specific conditions for reactivity should be optimized by the end user.  Expect a band approximately 12.5 kDa in size corresponding to reduced mature TGFb1 by western blotting.

Recommended Dilutions: ELISA: 1:40000; Western Blot: 1:100 - 1:500;

Type: Primary
Antigen: TGFB1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR