Anti-NEK2 Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK600-401-452EA 783 EUR
ROCK600-401-452
Anti-NEK2 Rabbit Polyclonal Antibody
Antikörper
This affinity purified antibody has been tested for use in ELISA and by western blot.  Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 52 kDa in size corresponding to NEK2 Protein by western blotting.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2000;

Type: Primary
Antigen: NEK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR