Anti-LAMB1 Rabbit Polyclonal Antibody (Biotin)

Lieferant: Rockland Immunochemicals
ROCK600-406-116EA 378 EUR
ROCK600-406-116
Anti-LAMB1 Rabbit Polyclonal Antibody (Biotin)
Antikörper
Anti-Laminin biotin conjugated antibody is suitable for western blotting, IHC, IP and for ELISA and for use with streptavidin reporter reagents. Researchers should determine optimal titers for applications that are not stated below.

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000; Immunoprecipitation: 1:100

Type: Primary
Antigen: LAMB1
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR