Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Rockland Immunochemicals
ROCK602-1502EA 173 EUR
ROCK602-1502
Anti-IgG (H+L) Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
Secondary Goat Anti-IgG (H&L) Reacts with Cat (Feline)

Recommended Dilutions: ELISA: 1:2000 - 1:6000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Cat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR