Anti-kappa light chain Goat Polyclonal Antibody (Biotin)

Lieferant: Rockland Immunochemicals
ROCK609-1610EA 188 EUR
ROCK609-1610
Anti-kappa light chain Goat Polyclonal Antibody (Biotin)
Antikörper
Anti-Human K antibody is suitable for ELISA, Western Blot and immunochemistry. Specific conditions for reactivity should be optimized by the end user.

Recommended Dilutions: ELISA: 1:200,000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: Igκ
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Light Chain
Host: Goat
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR