Anti-IgG (H+L) Goat Polyclonal Antibody (Peroxidase)

Lieferant: Rockland Immunochemicals
ROCK611-103-122EA 236 EUR
ROCK611-103-122
Anti-IgG (H+L) Goat Polyclonal Antibody (Peroxidase)
Antikörper
Secondary Goat Anti-IgG (H&L) Reacts with Rabbit (Lapine)

Recommended Dilutions: ELISA: 1:20000 - 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR