Anti-IgG F(ab')2 Goat Polyclonal Antibody (AP (Alkaline Phosphatase))

Lieferant: Rockland Immunochemicals
ROCK611-1504EA 188 EUR
ROCK611-1504
Anti-IgG F(ab')2 Goat Polyclonal Antibody (AP (Alkaline Phosphatase))
Antikörper
This product has been assayed against 1.0 µg of Rabbit IgG in a standard capture ELISA using pNPP p-nitrophenyl phosphate code # NPP-10 as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:500 - 1:2500; Immunochemistry: 1:200 - 1:1000;
Do not freeze!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope: F(ab')2
Host: Goat
Isotype: IgG
Reactivity: Rabbit
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR