Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (Texas Red®)

Lieferant: Rockland Immunochemicals
ROCK613-4930EA 245 EUR
ROCK613-4930
Anti-IgG F(ab')2 Rabbit Polyclonal Antibody (Texas Red®)
Antikörper
Secondary Rabbit Anti-F(ab')2 Reacts with Sheep (Ovine)

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 500 µl

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: F(ab')2
Host: Rabbit
Isotype: IgG
Reactivity: Sheep
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR