Anti-IgG (H+L) Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK614-4102EA 185 EUR
ROCK614-4102
Anti-IgG (H+L) Rabbit Polyclonal Antibody
Antikörper
Type: Secondary
Antigen: IgG (Pig) (pig IgG)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: none
Reactivity: Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR