Anti-IgA/IgG/IgM Goat Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK617-101-130EA 155 EUR
ROCK617-101-130
Anti-IgA/IgG/IgM Goat Polyclonal Antibody
Antikörper
Anti-Monkey IgG IgA IgM Antibody is suitable for immunoassays where reactivity to different classes of monkey immunoglobulins is desired. Antibody has been tested by ELISA, western blot immunoblot, and immunohistochemisty.

Recommended Dilutions: ELISA: 1:100000; Western Blot: 1:2000 - 1:10000; Immunochemistry: 1:1000 - 1:5000;

Type: Secondary
Antigen: IgA/IgG/IgM
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgA
Reactivity: Monkey
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR