Anti-Biotin Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: Rockland Immunochemicals
ROCK700-103-098EA 976 EUR
ROCK700-103-098
Anti-Biotin Goat Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Suitable for immunoblotting (western or dot blot), ELISA, immunoprecipitation and most immunological methods requiring high titer and specificity.

Recommended Dilutions: ELISA: 1:2000 - 1:10000; Western Blot: 1:200 - 1:1000; Immunochemistry: 1:100 - 1:500
Restore with deionized water (or equivalent); 1.0 ml
Do not add Sodium Azide!

Type: Tag
Antigen: Biotin
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Goat
Isotype: IgG
Reactivity: pan
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR