Anti-IgG (H+L) Goat Polyclonal Antibody (Texas Red®)

Lieferant: Rockland Immunochemicals
ROCK706-109-002EA 366 EUR
ROCK706-109-002
Anti-IgG (H+L) Goat Polyclonal Antibody (Texas Red®)
Antikörper
Secondary Goat Anti-IgG (H&L) Reacts with Guinea Pig

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Heavy and Light chain
Host: Goat
Isotype: IgG
Reactivity: Guinea Pig
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR