Anti-kappa light chain Goat Polyclonal Antibody (Texas Red®)

Lieferant: Rockland Immunochemicals
ROCK709-1910EA 210 EUR
ROCK709-1910
Anti-kappa light chain Goat Polyclonal Antibody (Texas Red®)
Antikörper
Secondary Goat Anti-κ kappa chain Reacts with Human

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000; Flow Cytometry: 1:500 - 1:2500
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: Igκ
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: Light Chain
Host: Goat
Isotype:
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR