Anti-IgM µ Goat Polyclonal Antibody (Texas Red®)

Lieferant: Rockland Immunochemicals
ROCK710-1907EA 243 EUR
ROCK710-1907
Anti-IgM µ Goat Polyclonal Antibody (Texas Red®)
Antikörper
Secondary Goat Anti-IgM (mu chain) Reacts with Mouse

Recommended Dilutions: FLISA: 1:10000 - 1:50000; Immunofluorescence: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgM µ
Clonality: Polyclonal
Clone:
Conjugation: Texas Red®
Epitope: µ-chain
Host: Goat
Isotype: IgG
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR