Anti-IgG (H+L) Rabbit Polyclonal Antibody (Peroxidase)

Lieferant: Rockland Immunochemicals
ROCK712-4326EA 235 EUR
ROCK712-4326
Anti-IgG (H+L) Rabbit Polyclonal Antibody (Peroxidase)
Antikörper
F(ab')2 Anti-Rat IgG (H&L) Antibody has been assayed against 1.0 µg of Rat IgG in a standard capture ELISA using ABTS (2,2’-azino-bis-(3-ethylbenthiazoline-6-sulfonic acid)) as a substrate for 30 minutes at room temperature.

Recommended Dilutions: ELISA: 1:10000 - 1:100000; Western Blot: 1:1000 - 1:10000; Immunochemistry: 1:500 - 1:2500;
Restore with deionized water (or equivalent); 500 µl
Do not add Sodium Azide!

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope: Heavy and Light chain
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR