Anti-IgG (H+L) Donkey Polyclonal Antibody (Biotin)

Lieferant: Rockland Immunochemicals
ROCK805-7602EA 401 EUR
ROCK805-7602
Anti-IgG (H+L) Donkey Polyclonal Antibody (Biotin)
Antikörper
Fab Anti-Goat IgG antibody is suitable for immunoblotting, ELISA, immunochemistry, immunomicroscopy as well as other antibody based assays using streptavidin or avidin conjugates requiring extremely low background levels, absence of F(c) mediated binding, lot-to-lot consistency, high titer and specificity.

Recommended Dilutions: ELISA: 1:600,000; Western Blot: 1:5000 - 1:20000; Immunochemistry: 1:1000 - 1:5000;
Restore with deionized water (or equivalent); 1.0 ml

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope: Heavy and Light chain
Host: Donkey
Isotype: IgG
Reactivity: Goat
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR