IgG2a Isotype Control (FITC (Fluorescein Isothiocyanate))

Lieferant: Rockland Immunochemicals
ROCK012-002-041EA 117 EUR
ROCK012-002-041
IgG2a Isotype Control (FITC (Fluorescein Isothiocyanate))
Antikörper
Rat IgG2a isotype control can be utilized as a control or standard reagent in Western Blotting, Flow Cytometry, and ELISA experiments where determination of sample isotype is important. Specific conditions should be optimized by user.

Recommended Dilutions: ELISA: 1:2000-1:20000; Flow Cytometry: 1:1000-1:5000;
Do not freeze!

Type: Control
Antigen: IgG2a
Clonality: Monoclonal
Clone:
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR