Anti-MMTV Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK100-401-P13EA 379 EUR
ROCK100-401-P13
Anti-MMTV Rabbit Polyclonal Antibody
Antikörper
Anti-MMTV Antibody has been tested for use in western blotting. Specific conditions for reactivity should be optimized by the end user. Expect band approximately 41.2 kDa in size corresponding to MMTV surface Protein by western blotting.

Recommended Dilutions: ELISA: 1:10000-1:20000; Western Blot: 1:1000-1:2000

Type: Primary
Antigen: MMTV
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Full length
Host: Rabbit
Isotype:
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR