Anti-PTPRC Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 25F6.F6.D8]

Lieferant: Rockland Immunochemicals
ROCK200-308-N86EA 527 EUR
ROCK200-308-N86
Anti-PTPRC Mouse Monoclonal Antibody (RPE (R-Phycoerythrin)) [clone: 25F6.F6.D8]
Antikörper
Anti-CD45.2 is useful for immunochemistry, Immunoprecipitation, and Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze! Light sensitive!

Type: Primary
Antigen: PTPRC
Clonality: Monoclonal
Clone: 25F6.F6.D8
Conjugation: RPE (R-Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR