Anti-IL2RA Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 3C7]

Lieferant: Rockland Immunochemicals
ROCK200-502-N82EA 438 EUR
ROCK200-502-N82
Anti-IL2RA Rat Monoclonal Antibody (FITC (Fluorescein isothiocyanate)) [clone: 3C7]
Antikörper
Anti-CD25 is useful for Flow Cytometry using mouse spleen cells, or an appropriate cell type (where indicated). Researchers should determine optimal titers for applications that are not stated.

Recommended Dilutions: Flow Cytometry: 10 µl/10^6 cells (0.1 µg);
Do not freeze!

Type: Primary
Antigen: IL2RA
Clonality: Monoclonal
Clone: 3C7
Conjugation: FITC (Fluorescein isothiocyanate)
Epitope:
Host: Rat
Isotype: IgG2b kappa
Reactivity: Mouse
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR