Anti-AKT2 Rat Monoclonal Antibody (Biotin) [clone: 16G11.E8]

Lieferant: Rockland Immunochemicals
ROCK200-506-E71EA 387 EUR
ROCK200-506-E71
Anti-AKT2 Rat Monoclonal Antibody (Biotin) [clone: 16G11.E8]
Antikörper
Anti-AKT2 BIOTIN Antibody is suitable for Flow Cytometry, ELISA, immunochemistry, and western blotting. Expect a band approximately 56 kDa in size corresponding to AKT2 Protein by western blotting in the appropriate cell lysate or extract.

Recommended Dilutions:
Restore with deionized water (or equivalent); 100 µl to 20 µl

Type: Primary
Antigen: AKT2
Clonality: Monoclonal
Clone: 16G11.E8
Conjugation: Biotin
Epitope: Center
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR