Anti-HIST2H3C Rabbit Polyclonal Antibody

Lieferant: Rockland Immunochemicals
ROCK600-401-P14EA 408 EUR
ROCK600-401-P14
Anti-HIST2H3C Rabbit Polyclonal Antibody
Antikörper
Anti-Histone H3 pT3/R2Me2s antibody is has been tested for use in Western Blot.

Recommended Dilutions: Immunofluorescence: 1:50-1:100; Western Blot: 1 µg/ml

Type: Primary
Antigen: HIST2H3C
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR