Anti-Chicken Serum Albumin Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI028983EA 472 EUR
USBI028983
Anti-Chicken Serum Albumin Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Chicken Serum Albumin (Chicken Serum Albumin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Chicken
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR