Anti-Pigeon Serum Albumin Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI029004EA 400 EUR
USBI029004
Anti-Pigeon Serum Albumin Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: Pigeon Serum Albumin (Pigeon Serum Albumin)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR