Anti-COX6A1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))

Lieferant: United States Biological
USBI029094EA 426 EUR
USBI029094
Anti-COX6A1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Antikörper
Type: Primary
Antigen: COX6A1 (cytochrome c oxidase subunit VIa polypeptide 1)
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR