Anti-EIF4A2 Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI029130EA 426 EUR
USBI029130
Anti-EIF4A2 Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: EIF4A2 (eukaryotic translation initiation factor 4A2)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR