Anti-GABARAP Rabbit Polyclonal Antibody
Lieferant: United States Biological
USBI029160EA 400 EUR
USBI029160
Anti-GABARAP Rabbit Polyclonal Antibody
Antikörper
Type: Primary
Antigen: GABARAP (GABA(A) receptor-associated protein)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human