Anti-HMGB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))

Lieferant: United States Biological
USBI029194EA 394 EUR
USBI029194
Anti-HMGB1 Rabbit Polyclonal Antibody (HRP (Horseradish Peroxidase))
Antikörper
Type: Primary
Antigen: HMGB1 (high mobility group box 1)
Clonality: Polyclonal
Clone:
Conjugation: HRP (Horseradish Peroxidase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR