Anti-LDHA Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI029198EA 394 EUR
USBI029198
Anti-LDHA Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 2mg/ml

Type: Primary
Antigen: LDHA (lactate dehydrogenase A)
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR