Anti-RAB1A Rabbit Polyclonal Antibody (Biotin)

Lieferant: United States Biological
USBI029285EA 426 EUR
USBI029285
Anti-RAB1A Rabbit Polyclonal Antibody (Biotin)
Antikörper
Type: Primary
Antigen: RAB1A (RAB1A, member RAS oncogene family)
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR