Anti-RuBisCO Rabbit Polyclonal Antibody

Lieferant: United States Biological
USBI029348EA 394 EUR
USBI029348
Anti-RuBisCO Rabbit Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 5 ?g/ml

Type: Primary
Antigen: RuBisCO
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity:
Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR